Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Aldrich Rhodium On Alumina
Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
1 g
5 g
25 g
Description
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.Rhodium on alumina (Rh/Al2O3) (5 wt. % loading, matrix alumina support, powder) is utilized as a catalyst for the steam reforming of ethanol for use in hydrogen production using molten carbonate fuel cells. Rh/Al2O3 can also be used as a catalyst in the production of hydrogen from sodium borohydride (NaBH4).
Specifications
Specifications
| MDL Number | MFCD00011201 |
| Quantity | 1 g |
| Synonym | Rh/Al2O3 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction