Learn More
Rhodium(III) chloride hydrate, Rh 38.0-45.5%
Description
Rhodium(III) chloride hydrate is used as a catalyst to prepare acetic acid. It is also used as a catalyst for reduction, oxidation of alkenes, hydration of acetylene and isomerization of alkenes. It is involved in the hydrosilylation of alfa,beta-unsaturated esters to form dimethylketene trimethylsilyl acetals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 100°C (decomposition) |
| Color | Red |
| Physical Form | Crystalline |
| Quantity | 1 g |
| Assay Percent Range | 38.5 to 45.5% |
| Solubility Information | Soluble in alcohol and water. Slightly soluble in acetone. |
| Formula Weight | 209.26 (Anhydrous) |
| Sensitivity | Hygroscopic |
| Appearance | Dark red crystals |
| Chemical Name or Material | Rhodium(III) chloride hydrate, Rh 38.5-45.5% |
Safety and Handling
| Hazard Category | H290-H302-H318-H341 |
| Hazard Statement | In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. This material and/or its container must be disposed of as hazardous waste. |
| Precautionary Statement | P201-P202-P234-P264b-P270-P281-P301+P312-P305+P351+P338-P308+P313-P310-P330-P390-P501c |
| DOTInformation | Hazard Class: 8; Packaging Group: III |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.