Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rhodium(III) nitrate, solution, ca. 10% (w/v) Rh in 20-25 wt% HNO3

Catalog No. AC318980100
Click to view available options
Quantity:
2 mL
10 mL
Packaging:
Glass bottle

CAS: 10139-58-9 | N3O9Rh | 288.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10139-58-9, 7697-37-2, 7732-18-5
Molecular Formula N3O9Rh
Molecular Weight (g/mol) 288.92
MDL Number MFCD00011202
InChI Key VXNYVYJABGOSBX-UHFFFAOYSA-N
Synonym rhodium nitrate, rhodium iii nitrate, rhodium trinitrate, nitric acid, rhodium 3+ salt, rhodium 3+ trinitrate, nitric acid, rhodium 3+ salt 3:1, nitric acid,rhodium 3+ salt 3:1, rhodiumtrinitrate, rhodium +3 nitrate, acmc-20ajsk
PubChem CID 150190
IUPAC Name rhodium(3+);trinitrate
SMILES [Rh+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Color Brown-Orange to Brown-Red
Physical Form Solution
Quantity 10 mL
Linear Formula Rh(NO3)3
Formula Weight 288.92
Percent Purity 20 to 25% (HNO3) (w/w), 10 to 14% (Rh) (w/v)
Packaging Glass bottle
Specific Gravity 1.29
Density 1.2900g/mL
Chemical Name or Material Rhodium(III) nitrate, Solution, ca.10% (w/w) Rh in 20-25 weight% HNO3
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May intensify fire; oxidizer.
May be corrosive to metals.
Harmful if swallowed.
May cause an allergic skin reaction.
Suspected of causing genetic defects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take any precaution to avoid mixing with combustibles.
In case of major
EINECSNumber 233-397-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.