Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rhodizonic Acid Dihydrate, Thermo Scientific™

Catalog No. AC157720050
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC157720050 5 g
AC157720010 1 g
AC157720100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC157720050 Supplier Thermo Scientific Chemicals Supplier No. 157720050

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118-76-3
Molecular Formula C6H2O6
Molecular Weight (g/mol) 170.076
InChI Key WCJLIWFWHPOTAC-UHFFFAOYSA-N
Synonym rhodizonic acid, 5,6-dihydroxycyclohex-5-ene-1,2,3,4-tetraone, 5,6-dihydroxy-5-cyclohexene-1,2,3,4-tetrone, dioxydiquinone, rhodizonsaure, dihydroxydiquinoyl, 16833-52-6 barium salt, 523-21-7 hydrochloride salt, 13021-40-4 dipotassium salt, 5-cyclohexene-1,2,3,4-tetrone, 5,6-dihydroxy
PubChem CID 67050
IUPAC Name 5,6-dihydroxycyclohex-5-ene-1,2,3,4-tetrone
SMILES C1(=C(C(=O)C(=O)C(=O)C1=O)O)O

Specifications

Color Brown
Infrared Spectrum Authentic
Quantity 5 g
Beilstein 08, 535
Formula Weight 206.11
Physical Form Fine Crystalline Powder
Chemical Name or Material Rhodizonic acid dihydrate
EINECSNumber 204-276-5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.