Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rhodopsin Antibody (4D2), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259690FR
Description
Rhodopsin Monoclonal specifically detects Rhodopsin in Amphibian, Avian, Fish, Mammal, Zebrafish samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
Rhodopsin | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
RHO | |
Bovine Rhodopsin | |
100 μL | |
GPCR, Phospho Specific, Signal Transduction, Vision | |
6010 | |
Amphibian, Avian, Fish, Mammalia, Zebrafish | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
4D2 | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen | |
CSNBAD1, MGC138309, OPN2MGC138311, opsin 2, rod pigment, opsin-2, retinitis pigmentosa 4, autosomal dominant, rhodopsin, RP4 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Binds specifically to the N-terminus of Rhodopsin. Does not detect Rhodopsin in invertebrates. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction