Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rhodopsin Antibody (B630), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP225160B
Description
Rhodopsin Monoclonal specifically detects Rhodopsin in Human, Mouse, Rat, Porcine, Bovine, Canine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Rhodopsin | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
RHO | |
Purified rhodopsin from bovine retina. | |
0.1 mL | |
GPCR, Phospho Specific, Signal Transduction, Vision | |
6010 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
B630 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
CSNBAD1, MGC138309, OPN2MGC138311, opsin 2, rod pigment, opsin-2, retinitis pigmentosa 4, autosomal dominant, rhodopsin, RP4 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Pig, Bovine, Canine, Equine | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction