Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rhodopsin Antibody (Rho 1D4), Janelia Fluor™ 646, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP281059JF646
Description
Rhodopsin Monoclonal specifically detects Rhodopsin in Human, Mouse, Rat, Amphibian, Bovine, Zebrafish samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Rhodopsin | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
RHO | |
Bleached bovine rod outer segment (ROS). | |
0.1 mL | |
GPCR, Phospho Specific, Signal Transduction, Vision | |
6010 | |
Human, Mouse, Rat, Amphibian, Bovine, Zebrafish | |
Purified |
Immunohistochemistry, Immunofluorescence | |
Rho 1D4 | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
CSNBAD1, MGC138309, OPN2MGC138311, opsin 2, rod pigment, opsin-2, retinitis pigmentosa 4, autosomal dominant, rhodopsin, RP4 | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
The antibody binds to the C-terminus of rhodopsin on the cytoplasmic side of disk membranes. The epitope has the following amino acid sequence: 'TETSQVAPA'. A limitation of the 1D4 tag, however, is that it has to be placed at the C-terminus of a protein. This is because the Rho1D4 monoclonal antibody requires a free carboxylate group for high affinity binding. Amidation of the carboxyl group lowers its immunoreactivity to the Rho1D4 antibody by over 100 fold. | |
Store at 4°C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction