Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rhodopsin Chimeric Recombinant Mouse Monoclonal Antibody (Rho 1D4)
GREENER_CHOICE

Catalog No. PIMA547924
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547924 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547924 Supplier Invitrogen™ Supplier No. MA547924
Add to Cart
Add to Cart

Mouse Recombinant Monoclonal Antibody

Specificity: The antibody binds to the C-terminus of rhodopsin on the cytoplasmic side of disk membranes. The epitope has the following amino acid sequence: 'TETSQVAPA'. A limitation of the 1D4 tag, however, is that it has to be placed at the C-terminus of a protein. This is because the Rho1D4 monoclonal antibody requires a free carboxylate group for high affinity binding. Amidation of the carboxyl group lowers its immunoreactivity to the Rho1D4 antibody by over 100 fold.

Visual pigments such as rhodopsin and porphyropsin are light-absorbing molecules that mediate vision. Rhodopsin consists of an apoprotein, opsin, covalently linked to 11-cis-retinal. This receptor is coupled to the activation of phospholipase C. Porphyropsin consists of opsin covalently linked to 11-cis 3,4-didehydroretinal.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rhodopsin Chimeric
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Radioimmune Assays (RIA), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone Rho 1D4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene RHO
Gene Accession No. P02699, P08100, P15409, P35359, P51489
Gene Alias CSNBAD1; L opsin; Long Wavelength Sensitive opsin; LWS opsin; MGC138309; MGC138311; Noerg1; OPN 2; Opn2; Ops; opsin 2; opsin 2, rod pigment; opsin-2; Red Opsin; retinal rod opsin pigment rh1.1; Rh; RH1; rh1.1; rho; rho protein; rhodopsin; rhodopsin (opsin 2, rod pigment) (retinitis pigmentosa 4, autosomal dominant); Rhodopsin (retinitis pigmentosa 4, autosomal dominant); rhodopsin-like-1.1 rod opsin; Rod Opsin; RP 4; RP4; seven transmembrane light receptor; unknown opsin-like protein; wu:fi06d11; zfo2; zfrho
Gene Symbols RHO
Host Species Mouse
Immunogen Bleached bovine rod outer segment (ROS).
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 212541, 24717, 30295, 509933, 6010
Target Species Human, Mouse, Rat, Bovine, Amphibian, Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.