Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rhodopsin Monoclonal Antibody (1D4), FITC
GREENER_CHOICE

Catalog No. PIMA545248
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545248 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545248 Supplier Invitrogen™ Supplier No. MA545248
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

1 μg/mL of MA5-45248 was sufficient for detection of rhodopsin in 10 μg of rat eye lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 40kDa. Recognizes Rhodopsin (native and recombinant forms). No known reactivity to other proteins. Binds specifically to the C-terminal epitope -T-E-T-S-Q-V-A-P-A-(COOH).

Visual pigments such as rhodopsin and porphyropsin are light-absorbing molecules that mediate vision. Rhodopsin consists of an apoprotein, opsin, covalently linked to 11-cis-retinal. This receptor is coupled to the activation of phospholipase C. Porphyropsin consists of opsin covalently linked to 11-cis 3,4-didehydroretinal.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rhodopsin
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 1D4
Concentration 1 mg/mL
Conjugate FITC
Formulation PBS, 9.1mM sodium bicarbonate with 640.91mM DMSO, 136.36mM ethanolamine and no preservative; pH 7.4
Gene RHO
Gene Accession No. P08100, P15409
Gene Alias CSNBAD1; L opsin; Long Wavelength Sensitive opsin; LWS opsin; MGC138309; MGC138311; Noerg1; OPN 2; Opn2; Ops; opsin 2; opsin 2, rod pigment; opsin-2; Red Opsin; retinal rod opsin pigment rh1.1; Rh; RH1; rh1.1; rho; rho protein; rhodopsin; rhodopsin (opsin 2, rod pigment) (retinitis pigmentosa 4, autosomal dominant); Rhodopsin (retinitis pigmentosa 4, autosomal dominant); rhodopsin-like-1.1 rod opsin; Rod Opsin; RP 4; RP4; seven transmembrane light receptor; unknown opsin-like protein; wu:fi06d11; zfo2; zfrho
Gene Symbols RHO
Host Species Mouse
Immunogen Bovine Rhodopsin.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 212541, 6010
Target Species Human, Mouse
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.