Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rhodopsin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572238
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572238 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572238 Supplier Invitrogen™ Supplier No. PA572238
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Rhodopsin Polyclonal Antibody for Western Blot

Visual pigments such as rhodopsin and porphyropsin are light-absorbing molecules that mediate vision. Rhodopsin consists of an apoprotein, opsin, covalently linked to 11-cis-retinal. This receptor is coupled to the activation of phospholipase C. Porphyropsin consists of opsin covalently linked to 11-cis 3,4-didehydroretinal.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rhodopsin
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene RHO
Gene Accession No. P35359
Gene Alias CSNBAD1; L opsin; Long Wavelength Sensitive opsin; LWS opsin; MGC138309; MGC138311; Noerg1; OPN 2; Opn2; Ops; opsin 2; opsin 2, rod pigment; opsin-2; Red Opsin; retinal rod opsin pigment rh1.1; Rh; RH1; rh1.1; rho; rho protein; rhodopsin; rhodopsin (opsin 2, rod pigment) (retinitis pigmentosa 4, autosomal dominant); Rhodopsin (retinitis pigmentosa 4, autosomal dominant); rhodopsin-like-1.1 rod opsin; Rod Opsin; RP 4; RP4; seven transmembrane light receptor; unknown opsin-like protein; wu:fi06d11; zfo2; zfrho
Gene Symbols RHO
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 62-93 amino acids from the human region of DANRE rho.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30295
Target Species Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.