Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rhodopsin Polyclonal Antibody, Invitrogen™

Catalog No. PIPA585608
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585608 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585608 Supplier Thermo Scientific Supplier No. PA585608

Rabbit Polyclonal Antibody

Keep as concentrated solution. Predicted reactivity: Dog (100%), Cat (100%), Pig (100%), Rabbit (100%), Guinea pig (100%). Positive Control: mouse eye. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Visual pigments such as rhodopsin and porphyropsin are light-absorbing molecules that mediate vision. Rhodopsin consists of an apoprotein, opsin, covalently linked to 11-cis-retinal. This receptor is coupled to the activation of phospholipase C. Porphyropsin consists of opsin covalently linked to 11-cis 3,4-didehydroretinal.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rhodopsin
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative, pH 7
Gene RHO
Gene Accession No. P08100, P15409, P51489
Gene Alias CSNBAD1, L opsin, Long Wavelength Sensitive opsin, LWS opsin, MGC138309, MGC138311, Noerg1, OPN 2, Opn2, Ops, opsin 2, opsin 2, rod pigment, opsin-2, Red Opsin, retinal rod opsin pigment rh1.1, Rh, RH1, rh1.1, rho, rho protein, rhodopsin, rhodopsin (opsin 2, rod pigment) (retinitis pigmentosa 4, autosomal dominant), Rhodopsin (retinitis pigmentosa 4, autosomal dominant), rhodopsin-like-1.1 rod opsin, Rod Opsin, RP 4, RP4, seven transmembrane light receptor, unknown opsin-like protein, wu:fi06d11, zfo2, zfrho
Gene Symbols RHO
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human Rhodopsin.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 212541, 24717, 6010
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.