Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RhoF Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298649AF700
Description
RhoF Polyclonal specifically detects RhoF in Human samples. It is validated for Western Blot.Specifications
RhoF | |
Polyclonal | |
Western Blot | |
FLJ20247, ras homolog gene family, member F (in filopodia), Rho family GTPase Rif, Rho in filopodia, rho-related GTP-binding protein RhoF, RIFARHF | |
Produced in rabbits immunized with E. coli-derived Human RhoF fragment. | |
0.1 mL | |
Signal Transduction | |
54509 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction