Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RhoF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584989
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584989 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584989 Supplier Invitrogen™ Supplier No. PA584989
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LLMVYSQGSF PEHYAPSVFE KYTASVTVGS KEVTLNLYD Highest antigen sequence indentity to the following orthologs: Mouse - 95%, Rat - 95%.

Plasma membrane-associated small GTPase which cycles between an active GTP-bound and an inactive GDP-bound state. Causes the formation of thin, actin-rich surface projections called filopodia. Functions cooperatively with CDC42 and Rac to generate additional structures, increasing the diversity of actin-based morphology.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RhoF
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RHOF
Gene Accession No. Q9HBH0
Gene Alias AI845056; Arhf; AV026554; Ifld1; induced in fatty liver dystrophy 1; ras homolog family member F (in filopodia); ras homolog family member F, filopodia associated; ras homolog gene family, member f; ras homolog gene family, member f (in filopodia); rho family GTPase Rif; Rho in filopodia; RHOF; rho-related GTP-binding protein RhoF; RIF
Gene Symbols RHOF
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RHOF. Recombinant protein control fragment (Product #RP-108276).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54509
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.