Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RhoG Polyclonal Antibody
RhoG Polyclonal Antibody
Supplier: Thermo Scientific PA558958
Description
RhoG Polyclonal Antibody for Western Blot, IHC

Specifications
RhoG | |
Polyclonal | |
Unconjugated | |
RHOG | |
2810426G09Rik; aplysia ras-related homolog G (RhoG); Arhg; ras homolog family member G; ras homolog G (RhoG); ras homolog gene family, member G; ras homolog gene family, member G (rho G); Rho G; Rhog; rho-related GTP-binding protein RhoG; Sid 10750; Sid10750 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
391 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P84095 | |
RHOG | |
Recombinant protein corresponding to Human RhoG. Recombinant protein control fragment (Product #RP-97508). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction