Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RhoG Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599704
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599704 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599704 Supplier Thermo Scientific Supplier No. PA599704
Add to Cart
Add to Cart

RhoG Polyclonal Antibody

RhoG Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen RhoG
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RHOG
Gene Accession No. P84095, P84096
Gene Alias 2810426G09Rik; aplysia ras-related homolog G (RhoG); Arhg; ras homolog family member G; ras homolog G (RhoG); ras homolog gene family, member G; ras homolog gene family, member G (rho G); Rho G; Rhog; rho-related GTP-binding protein RhoG; Sid 10750; Sid10750
Gene Symbols RHOG
Host Species Rabbit
Immunogen A synthesized peptide derived from human RHOG(Accession P84095), corresponding to amino acid residues Y64-G114.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 391, 56212
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.