Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RhoGAP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA521485
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA521485 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA521485 Supplier Invitrogen™ Supplier No. PA521485
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Jurkat, NT2D1. Predicted reactivity: Mouse (81%), Bovine (86%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the rhoGAP family of proteins which play a role in the regulation of small GTP-binding proteins belonging to the RAS superfamily. The protein encoded by the orthologous gene in rat is localized to the Golgi complex and can redistribute to microtubules. The rat protein stimulates the activity of some Rho GTPases in vitro. Genomic deletions of this gene and a neighboring gene have been found in patients with nephrogenic diabetes insipidus. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RhoGAP4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene ARHGAP4
Gene Accession No. P98171
Gene Alias A130009C12Rik; A530015A20Rik; ARHGAP4; C1; GTPase-activating protein p115; KIAA0131; mKIAA0131; p115; RG RhoGAP4; RGC1; Rho GTPase activating protein 4; rho GTPase-activating protein 4; Rho-GAP hematopoietic protein C1; RHOGAP4; rho-type GTPase-activating protein 4; SrGAP4
Gene Symbols ARHGAP4
Host Species Rabbit
Immunogen Recombinant fragment corresponding to a region within amino acids 1 and 280 of ARHGAP4 (Uniprot ID#P98171).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 393
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.