Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RhoGDI Antibody (2G3) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$560.50
Specifications
Antigen | RhoGDI |
---|---|
Clone | 2G3 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, CyTOF |
Description
RhoGDI Monoclonal specifically detects RhoGDI in Human, Mouse, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, CyTOF-ready.Specifications
RhoGDI | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, CyTOF | |
Unconjugated | |
RUO | |
Human, Mouse, Primate, Monkey | |
GDIA1, MGC117248, Rho GDI 1, Rho GDP dissociation inhibitor (GDI) alpha, rho GDP-dissociation inhibitor 1, RHOGDI, Rho-GDI alpha, RHOGDI-1 | |
ARHGDIA | |
IgG1 | |
Affinity Purified |
2G3 | |
Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, CyTOF-ready | |
Monoclonal | |
Mouse | |
Signal Transduction | |
PBS with 0.05% Sodium Azide | |
396 | |
Purified recombinant fragment of human ARHGDIA (AA: FULL(1-204)) expressed in E. Coli. | |
Primary | |
26 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title