Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RhoGDI Mouse anti-Human, Mouse, Rat, Clone: 1B1G9, Proteintech
SDP

Catalog No. 501736903 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6903 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6903 Supplier Proteintech Group Inc Supplier No. 664801IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Aplysia Ras-related homologs (ARHs), also called Rho genes, belong to the RAS gene superfamily encoding small guanine nucleotide exchange (GTP/GDP) factors. The ARH proteins may be kept in the inactive, GDP-bound state by interaction with GDP dissociation inhibitors, such as ARHGDIA.

Specifications

Antigen RhoGDI
Applications Western Blot
Classification Monoclonal
Clone 1B1G9
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene ARHGDIA
Gene Accession No. P52565, Q5XI73, Q99PT1
Gene Alias aGDI, ARHGDIA, GDIA1, Rho GDI 1, Rho GDI alpha, RHOGDI, RHOGDI 1
Gene Symbols Arhgdia
Host Species Mouse
Immunogen RhoGDI Fusion Protein Ag0791
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 192662, 360678, 396
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.