Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RhoGDI Rabbit anti-Human, Mouse, Rat, Monkey, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$243.50 - $524.00
Specifications
Antigen | RhoGDI |
---|---|
Applications | Western Blot, Immunofluorescence, KnockDown |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
RhoGDI Polyclonal specifically detects RhoGDI in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Knockout Validated.Specifications
RhoGDI | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat, Monkey | |
GDIA1, MGC117248, Rho GDI 1, Rho GDP dissociation inhibitor (GDI) alpha, rho GDP-dissociation inhibitor 1, RHOGDI, Rho-GDI alpha, RHOGDI-1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-204 of human RhoGDI (NP_001172006.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, Immunofluorescence, KnockDown | |
Unconjugated | |
Rabbit | |
Signal Transduction | |
PBS with 50% glycerol, pH7.3. | |
396 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title