Learn More
RHOH Mouse anti-Human, Clone: OTI9F4, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809996

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene is a member of the Ras superfamily of guanosine triphosphate (GTP)-metabolizing enzymes. The encoded protein is expressed in hematopoietic cells, where it functions as a negative regulator of cell growth and survival. This gene may be hypermutated or misexpressed in leukemias and lymphomas. Chromosomal translocations in non-Hodgkin′s lymphoma occur between this locus and B-cell CLL/lymphoma 6 (BCL6) on chromosome 3, leading to the production of fusion transcripts. Alternative splicing in the 5′ untranslated region results in multiple transcript variants that encode the same protein.Specifications
RHOH | |
Monoclonal | |
Unconjugated | |
RHOH | |
ARHH, TTF | |
Mouse | |
Affinity Chromatography | |
RUO | |
399 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI9F4 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q15669 | |
RHOH | |
Human recombit protein fragment corresponding to amino acids 97-190 of human RHOH produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.