Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RhoH Polyclonal Antibody
RhoH Polyclonal Antibody
Supplier: Thermo Scientific PA575334
Description
RhoH Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

Specifications
RhoH | |
Polyclonal | |
Unconjugated | |
RHOH | |
5830400A04Rik; ARHH; AU019774; GTP-binding protein TTF; ras homolog family member H; ras homolog gene family, member H; RHOH; rho-related GTP-binding protein RhoH; Translocation three four protein; TTF; TTF, translocation three four | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
305341, 399, 74734 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q15669, Q9D3G9 | |
RHOH | |
Synthetic peptide corresponding to amino acids 102-152 of Human RhoH. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction