Learn More
Invitrogen™ RhoH Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA592920
Description
Immunogen sequence: MLSSIKCVLV GDSAVGKTSL LVRFTSETFP EAYKPTVYEN TGVDVFMDGI QISLGLWDTA GNDAFRSIRP LSYQQADVVL MCYSVANHNS FLNLKNKWIG EIRSNLPCTP VLVVATQTDQ REMGPHRASC VNAMEGKKLA QDVRAKGYLE CSALSNRGVQ QVFECAVRTA VNQARRRNRR RLFSINECKI F; Positive Samples: BT-474, SW620, Jurkat, HT-29, Mouse spleen, Mouse thymus, Mouse liver; Cellular Location: Cell membrane, Cytoplasm, Cytoplasmic side, Lipid-anchor.
The Rho subfamily of small GTP-binding proteins mediates many fundamental cellular functions. The commonly studied members (Rho, Rac, and CDC42) regulate actin reorganization and affect diverse cellular responses, including adhesion, cytokinesis, and motility. RhoH, also known as TTF (Translocation Three Four), Rho-related GTP-binding protein and ras homolog gene family member H, is unlike most other small G proteins. Most small G proteins are expressed ubiquitously, however, Rho H is expressed only in hemopoietic cells and tissues. Translocations and a high frequency of Rho H mutation have been detected in primary lymphoma cells. Rho H expression has also been observed in activated neutrophils. RhoH is GTPase deficient, remaining in a GTP-bound activated state without cycling. Rho H may be involved in the functional differentiation of T cells and in cytoskeleton organization. The RhoH/TTF (ARHH) gene maps to chromosome 4p13 and encodes a 191 -amino acid polypeptide.
Specifications
RhoH | |
Polyclonal | |
Unconjugated | |
RHOH | |
5830400A04Rik; ARHH; AU019774; GTP-binding protein TTF; ras homolog family member H; ras homolog gene family, member H; RHOH; rho-related GTP-binding protein RhoH; Translocation three four protein; TTF; TTF, translocation three four | |
Rabbit | |
Affinity Chromatography | |
RUO | |
399, 74734 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1.32 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q15669, Q9D3G9 | |
RHOH | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-191 of human RHOH (NP_0043011). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.