Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RhoJ Antibody (OTI1D7), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273892B
Description
RhoJ Monoclonal specifically detects RhoJ in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
RhoJ | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
RHOJ | |
Full length recombinant protein of human RhoJ (NP_065714) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI1D7 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
ARHJTC10B, FLJ14445, ras homolog gene family, member J, RASL7B, Ras-like protein family member 7B, RAS-like, family 7, member B, RHOI, rho-related GTP-binding protein RhoJ, Tc10-like GTP-binding protein, TC10-like Rho GTPase, TCLMGC34777 | |
Mouse | |
Affinity Purified | |
Signal Transduction | |
57381.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction