Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RhoJ Monoclonal Antibody (OTI1D7)
GREENER_CHOICE

Catalog No. PIMA526170
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526170 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526170 Supplier Invitrogen™ Supplier No. MA526170
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RhoJ Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

ARHJ belongs to the Rho family of small GTP-binding proteins. Rho proteins regulate the dynamic assembly of cytoskeletal components for several physiologic processes, such as cell proliferation and motility and the establishment of cell polarity. They are also involved in pathophysiologic process, such as cell transformation and metastasis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RhoJ
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI1D7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RHOJ
Gene Accession No. Q9H4E5
Gene Alias 1110005O19Rik; Arhj; AW210585; FLJ14445; MGC34777; ras homolog family member J; ras homolog gene family, member J; RASL7B; ras-like protein family member 7B; RAS-like, family 7, member B; RHOI; RHOJ; Rho-related GTP-binding protein RhoJ; Rhot; si:dkey-100h21.1; TC10B; Tc10l; tc10-like GTP-binding protein; tc10-like GTP-binding protein TCL; TC10-like Rho GTPase; TCL
Gene Symbols RHOJ
Host Species Mouse
Immunogen Full length human recombinant protein of RHOJ produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57381
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.