Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RhoJ Monoclonal Antibody (OTI2C10)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA526142
Description
RhoJ Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
ARHJ belongs to the Rho family of small GTP-binding proteins. Rho proteins regulate the dynamic assembly of cytoskeletal components for several physiologic processes, such as cell proliferation and motility and the establishment of cell polarity. They are also involved in pathophysiologic process, such as cell transformation and metastasis.
Specifications
RhoJ | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9H4E5 | |
RHOJ | |
Full length human recombinant protein of RHOJ produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
OTI2C10 | |
Unconjugated | |
RHOJ | |
1110005O19Rik; Arhj; AW210585; FLJ14445; MGC34777; ras homolog family member J; ras homolog gene family, member J; RASL7B; ras-like protein family member 7B; RAS-like, family 7, member B; RHOI; RHOJ; Rho-related GTP-binding protein RhoJ; Rhot; si:dkey-100h21.1; TC10B; Tc10l; tc10-like GTP-binding protein; tc10-like GTP-binding protein TCL; TC10-like Rho GTPase; TCL | |
Mouse | |
Affinity chromatography | |
RUO | |
57381 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction