Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RhoJ Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA544776
Description
Peptide sequence: LGLYDTAGQE DYNQLRPLSY PNTDVFLICF SVVNPASYHN VQEEWVPELK Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 86%; Zebrafish: 100%.
ARHJ belongs to the Rho family of small GTP-binding proteins. Rho proteins regulate the dynamic assembly of cytoskeletal components for several physiologic processes, such as cell proliferation and motility and the establishment of cell polarity. They are also involved in pathophysiologic process, such as cell transformation and metastasis.
Specifications
| RhoJ | |
| Polyclonal | |
| Unconjugated | |
| RHOJ | |
| 1110005O19Rik; Arhj; AW210585; FLJ14445; MGC34777; ras homolog family member J; ras homolog gene family, member J; RASL7B; ras-like protein family member 7B; RAS-like, family 7, member B; RHOI; RHOJ; Rho-related GTP-binding protein RhoJ; Rhot; si:dkey-100h21.1; TC10B; Tc10l; tc10-like GTP-binding protein; tc10-like GTP-binding protein TCL; TC10-like Rho GTPase; TCL | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 57381 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9H4E5 | |
| RHOJ | |
| Synthetic peptide directed towards the middle region of human RHOJ. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction