Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Rhosin hydrochloride

Catalog No. 500310R
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-031-0R 10 mg
50-035-0R 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 50-031-0R Supplier Tocris Bioscience™ Supplier No. 5003/10
This item is not returnable. View return policy

Rho GTPase inhibitor

Rhosin hydrochloride is a Rho GTPase inhibitor (Kd = 354 nM). Inhibits binding of RhoA to guanine nucleotide exchange factors (GEFs) including LARG, DBL, LBC, p115 RhoGEF and PDZ RhoGEF. Has no effect on the binding of Rac1 or Cdc42 to their respective GEFs. Suppresses RhoA/C-mediated breast cancer cell migration and invasion in vitro. Also induces neurite outgrowth in synergy with NGF.

Specifications

Quantity 10 mg
Product Type Rhosin hydrochloride
Purity >98%
Molecular Weight (g/mol) 394.86
Inhibitors Rho GTPase Inhibitors

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.