Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rhot1 Antibody (CL1095), Novus Biologicals™
SDP

Catalog No. NB395327 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB395327 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB395327 Supplier Novus Biologicals Supplier No. NBP25296525UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Rhot1 Monoclonal antibody specifically detects Rhot1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen Rhot1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL1095
Conjugate Unconjugated
Dilution Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS (pH 7.2), 40% Glycerol
Gene Alias ARHT1, EC 3.6.5, EC 3.6.5.-, FLJ11040, FLJ12633, hMiro-1, MIRO-1mitochondrial Rho GTPase 1, mitochondrial Rho 1, rac-GTP binding protein-like protein, Rac-GTP-binding protein-like protein, Ras homolog gene family member T1, ras homolog gene family, member T1
Host Species Mouse
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: THIVDYSEAEQSDEQLHQEISQANVICIVYAVNNKHSIDKVTSRWIPLINERTDKDSRLPLILVGNKSDLVEYSSMETILPIMNQYTEIE
Purification Method Protein A purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 55288
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.