Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rhot1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 5 publications
Supplier: Novus Biologicals NBP159021
Description
Rhot1 Polyclonal specifically detects Rhot1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
Rhot1 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
ARHT1, EC 3.6.5, EC 3.6.5.-, FLJ11040, FLJ12633, hMiro-1, MIRO-1mitochondrial Rho GTPase 1, mitochondrial Rho 1, rac-GTP binding protein-like protein, Rac-GTP-binding protein-like protein, Ras homolog gene family member T1, ras homolog gene family, member T1 | |
Rabbit | |
71 kDa | |
100 μL | |
Signal Transduction | |
55288 | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
IgG |
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunoprecipitation, Immunohistochemistry-Paraffin 4-8 ug/ml | |
Q8IXI2 | |
RHOT1 | |
Synthetic peptides corresponding to RHOT1(ras homolog gene family, member T1) The peptide sequence was selected from the N terminal of RHOT1 (NP_060777). Peptide sequence MKKDVRILLVGEPRVGKTSLIMSLVSEEFPEEVPPRAEEITIPADVTPER. | |
Affinity purified | |
RUO | |
Primary | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction