Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RHOT1 Recombinant Rabbit Monoclonal Antibody (6B8R1)
GREENER_CHOICE

Catalog No. FEMA551305
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA551305 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA551305 Supplier Invitrogen™ Supplier No. MA551305
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: QSTFDKHDLD RDCALSPDEL KDLFKVFPYI PWGPDVNNTV CTNERGWITY QGFLSQWTLT TYLDVQRCLE YLGYLGYSIL TEQESQASAV TVTRDKKIDL QKKQTQRNVF RCNVIGVKNC GKSGVLQALL GRNLMRQKKI REDHKSYYAI NTVYVYGQEK YLLLHDISES EFLTEAEIIC DVVCLVYDVS NPKSFEYCAR IFKQHFMDSR IPCLIVAAKS DLHEVKQEYS ISPTDFCRKH KMPPPQAFTC NTADAPSKDI FVKLTTMAMY PHVTQADLKS S.

Mitochondrial GTPase involved in mitochondrial trafficking. RHOT1 is probably involved in control of anterograde transport of mitochondria and their subcellular distribution.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RHOT1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone 6B8R1
Concentration 0.237 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene RHOT1
Gene Accession No. Q8BG51, Q8IXI2
Gene Alias 2210403N23Rik; AA415293; AF244542; AI834919; Arht1; C430039G08Rik; LOW QUALITY PROTEIN: mitochondrial Rho GTPase 1; MGC128572 protein; Miro1; MIRO-1; mitochondrial Rho (MIRO) GTPase 1; mitochondrial Rho GTPase 1; rac-GTP binding protein-like protein; Rac-GTP-binding protein-like protein; ras homolog family member T1; Ras homolog gene family member T1; ras homolog gene family, member T1; Rhot1
Gene Symbols RHOT1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 310-590 of human RHOT1/MIRO1 (NP_060777.3).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 303351, 55288, 59040
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.