Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RHOT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100317
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100317 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100317 Supplier Invitrogen™ Supplier No. PA5100317
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total RHOT2.

This gene encodes a member of the Rho family of GTPases. The encoded protein is localized to the outer mitochondrial membrane and plays a role in mitochondrial trafficking and fusion-fission dynamics.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RHOT2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RHOT2
Gene Accession No. Q8IXI1, Q8JZN7
Gene Alias Arht2; C16orf39; hypothetical protein LOC557574; Miro2; MIRO-2; mitochondrial Rho (MIRO) GTPase 2; mitochondrial Rho 2; mitochondrial Rho GTPase 2; ras homolog family member T2; Ras homolog gene family member T2; ras homolog gene family, member T2; RASL; Rhot2; zgc:123249
Gene Symbols RHOT2
Host Species Rabbit
Immunogen A synthesized peptide derived from human RHOT2(Accession Q8IXI1), corresponding to amino acid residues R191-V241.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 214952, 89941
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.