Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RHOT2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA545305
Description
Peptide sequence: TIPADVTPEK VPTHIVDYSE AEQTDEELRE EIHKANVVCV VYDVSEEATI Sequence homology: Dog: 86%; Guinea Pig: 79%; Horse: 79%; Human: 100%; Mouse: 86%; Rat: 86%; Zebrafish: 91%.
This gene encodes a member of the Rho family of GTPases. The encoded protein is localized to the outer mitochondrial membrane and plays a role in mitochondrial trafficking and fusion-fission dynamics.
Specifications
| RHOT2 | |
| Polyclonal | |
| Unconjugated | |
| RHOT2 | |
| Arht2; C16orf39; hypothetical protein LOC557574; Miro2; MIRO-2; mitochondrial Rho (MIRO) GTPase 2; mitochondrial Rho 2; mitochondrial Rho GTPase 2; ras homolog family member T2; Ras homolog gene family member T2; ras homolog gene family, member T2; RASL; Rhot2; zgc:123249 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 89941 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8IXI1 | |
| RHOT2 | |
| Synthetic peptide directed towards the N-terminal of human RHOT2 (aa 41-90). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction