Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RHOT2 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA552960
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552960 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552960 Supplier Thermo Scientific Supplier No. PA552960
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RSCLGHLGYL GYPTLCEQDQ AHAITVTREK RLDQEKGQTQ RSVLLCKVVG ACGVGKSAFL QAFLGRGLGH QDTREQPPGY AIDTVQVNGQ EKYLILC Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 75%.

This gene encodes a member of the Rho family of GTPases. The encoded protein is localized to the outer mitochondrial membrane and plays a role in mitochondrial trafficking and fusion-fission dynamics.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RHOT2
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RHOT2
Gene Accession No. Q8IXI1
Gene Alias Arht2; C16orf39; hypothetical protein LOC557574; Miro2; MIRO-2; mitochondrial Rho (MIRO) GTPase 2; mitochondrial Rho 2; mitochondrial Rho GTPase 2; ras homolog family member T2; Ras homolog gene family member T2; ras homolog gene family, member T2; RASL; Rhot2; zgc:123249
Gene Symbols RHOT2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RHOT2
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89941
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.