Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RhoV Antibody - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. p-7199907 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018494 0.1 mg
NBP130656 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018494 Supplier Novus Biologicals Supplier No. NBP1306560.1MG
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RhoV Polyclonal specifically detects RhoV in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen RhoV
Applications Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Formulation Sterilized water with No Preservative
Gene Alias ARHV, CDC42-like GTPase 2, CHP, GTP-binding protein-like 2, ras homolog gene family, member V, Rho GTPase-like protein ARHV, rho-related GTP-binding protein RhoV, Wnt-1 regulated Cdc42 homolog 2, Wnt-1 responsive Cdc42 homolog 2, WRCH1-related GTPase, WRCH-2, WRCH2Chp
Gene Symbols RHOV
Host Species Rabbit
Immunogen A synthetic peptide from the c-terminal region of human RHOV (CDC42-like GTPase 2) conjugated to an immunogenic carrier protein was used as the immunogen. The antigen is homologous in rat and mouse.
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171177
Test Specificity Appears to be specific for RhoV.
Reconstitution Reconstitute with 0.5 ml sterilized water.
Target Species Rat, Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.