Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    Invitrogen™ RhoV Polyclonal Antibody
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551217
Description
The antibody detects endogenous levels of total RHOV protein.
The Rho subfamily of Ras-related GTPases controls multiple aspects of cell function, including cytoskeletal rearrangement, nuclear signaling and cell growth. Rho V (ras homolog gene family, member V), also known as Rho GTPase-like protein ARHV, CHP or WRCH2 (Wnt-1 responsive Cdc42 homolog 2), is a 236 amino acid protein that controls the actin cytoskeleton through activation of the JNK pathway. A member of the Rho family and small GTPase superfamily, Rho V functions as a lipid anchor at the cytoplasmic side of the cell membrane and is expressed in placenta, pancreas and fetal brain. Rho V is implicated in cell transformation and is encoded by a gene located on human chromosome 15, which houses over 700 genes and comprises nearly 3% of the human genome. Angelman syndrome, Prader-Willi syndrome, Tay-Sachs disease and Marfan syndrome are all associated with defects in chromosome 15-localized genes.
            Specifications
| RhoV | |
| Polyclonal | |
| Unconjugated | |
| Rhov | |
| A030005A06Rik; ARHV; calcium binding protein p22; CDC42-like GTPase 2; CHP; GTP-binding protein-like 2; ras homolog family member V; ras homolog gene family, member V; RGD:628824}; rho family GTPase Chp; Rho GTPase-like protein ARHV; rho-related GTP-binding protein RhoV; RHOV; rhov {ECO:0000312; Wnt-1 regulated Cdc42 homolog 2; Wnt-1 responsive Cdc42 homolog 2; WRCH1-related GTPase; WRCH2; WRCH-2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 171177 | |
| -20°C | |
| Liquid | 
| Immunohistochemistry (Paraffin) | |
| 0.9 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q96L33 | |
| Rhov | |
| Full length fusion protein of human RHOV. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction