Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RhoV Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562781
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562781 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562781 Supplier Invitrogen™ Supplier No. PA562781
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KEVFDSAILS AIEHKARLEK KLNAKGVRTL SRCRWKKFFC FV Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

The Rho subfamily of Ras-related GTPases controls multiple aspects of cell function, including cytoskeletal rearrangement, nuclear signaling and cell growth. Rho V (ras homolog gene family, member V), also known as Rho GTPase-like protein ARHV, CHP or WRCH2 (Wnt-1 responsive Cdc42 homolog 2), is a 236 amino acid protein that controls the actin cytoskeleton through activation of the JNK pathway. A member of the Rho family and small GTPase superfamily, Rho V functions as a lipid anchor at the cytoplasmic side of the cell membrane and is expressed in placenta, pancreas and fetal brain. Rho V is implicated in cell transformation and is encoded by a gene located on human chromosome 15, which houses over 700 genes and comprises nearly 3% of the human genome. Angelman syndrome, Prader-Willi syndrome, Tay-Sachs disease and Marfan syndrome are all associated with defects in chromosome 15-localized genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RhoV
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Rhov
Gene Accession No. Q96L33
Gene Alias A030005A06Rik; ARHV; calcium binding protein p22; CDC42-like GTPase 2; CHP; GTP-binding protein-like 2; ras homolog family member V; ras homolog gene family, member V; RGD:628824}; rho family GTPase Chp; Rho GTPase-like protein ARHV; rho-related GTP-binding protein RhoV; RHOV; rhov {ECO:0000312; Wnt-1 regulated Cdc42 homolog 2; Wnt-1 responsive Cdc42 homolog 2; WRCH1-related GTPase; WRCH2; WRCH-2
Gene Symbols Rhov
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RhoV. Recombinant protein control fragment (Product #RP-104211).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171177
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.