Learn More
Invitrogen™ Rhox11 Polyclonal Antibody

Description
Rhox11 Polyclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | Rhox11 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Whole serum with 50% glycerol and 0.025% sodium azide |
| Gene | Rhox11 |
| Gene Accession No. | 0 |
| Gene Alias | BC049711; Gm39; reproductive homeobox 11; reproductive homeobox on X chromosome, 11; Rhox11; RP23-322E15.4 |
| Gene Symbols | Rhox11 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.