Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rhox13 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA523371

Catalog No. PIPA523371


Only null left
Add to Cart

Description

Description

Rhox13 Polyclonal Antibody for Western Blot

The Rhox-13 protein is a reproductive homeobox protein found on the X chromosome. This transcription factor belongs to the homeobox gene family whose expression organizes programs of development. Reproductive homeobox genes regulate development of reproductive tissues including the testis, ovary, and placenta. Most of the Rhox genes are expressed in Sertoli cells, the nurse cells in direct contact with developing male germ cells. This suggests that they regulate the expression of somatic-cell gene products critical for germ cell development. Transcripts of this protein are found in the testis and ovary as early as 13.5 days post coitum. Transcription ceases in the ovary three days post partum, but continues in the testis through adulthood.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Rhox13
Polyclonal
Unconjugated
Rhox13
1700123J19Rik; Homeobox protein Rhox13; novel homeobox domain-containing protein; reproductive homeobox 13; Reproductive homeobox protein 13; Rhox13; RP23-322E15.7
Rabbit
Antigen affinity chromatography
RUO
73614
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
F6YCR7
Rhox13
A portion of amino acids 1-50 of Rhox-13.
100 μg
Primary
Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.