Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rhox9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA523134
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA523134 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA523134 Supplier Invitrogen™ Supplier No. PA523134
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence homology: rat (93%), human (72%), opossum (71%).

Reproductive homeobox on X chromosome 9, also known as Reproductive Homeobox (Rhox)protein GPBOX, is a nuclear protein product of a 12 X-linked novel homeobox gene family that plays an important role during postnatal testis development. The members of this family are characterized by a conserved 60-amino-acid DNA-binding homeodomain. These transcription factors not only play an important role in various developmental processes such as DNA-dependent transcriptional regulation, body-plan specification, pattern formation, and cell fate determination, but also in understanding the development and physiology of the human reproductive system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rhox9
Applications Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Whole serum with 50% glycerol and 0.025% sodium azide
Gene Rhox9
Gene Accession No. 0
Gene Alias 1600026O01Rik; Gpbox; placenta specific homeobox 2; Psx2; reproductive homeobox 9; reproductive homeobox on X chromosome, 9; Rhox9; RP23-202F23.5
Gene Symbols Rhox9
Host Species Rabbit
Immunogen Within the range of amino acids 82-112 of mouse Rhox-9 protein.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 104384
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.