Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RHOXF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563313
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563313 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563313 Supplier Invitrogen™ Supplier No. PA563313
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AMEGPQPENM QPRTRRTKFT LLQVEELESV FRHTQYPDVP TRRELAENLG VTEDKVRVWF Highest antigen sequence identity to the following orthologs: Mouse - 48%, Rat - 47%.

This gene is a member of the PEPP subfamily of paired-like homoebox genes. The gene may be regulated by androgens and epigenetic mechanisms. The encoded nuclear protein is likely a transcription factor that may play a role in human reproduction. [provided by RefSeq, Dec 2012].
TRUSTED_SUSTAINABILITY

Specifications

Antigen RHOXF1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RHOXF1
Gene Accession No. Q8NHV9
Gene Alias OTEX; ovary-, testis- and epididymis-expressed gene protein; Paired-like homeobox protein PEPP-1; PEPP subfamily gene 1; PEPP1; Rhox homeobox family member 1; Rhox homeobox family, member 1; RHOXF1; ZKSCAN21
Gene Symbols RHOXF1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RHOXF1. Recombinant protein control fragment (Product #RP-100915).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 158800
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.