Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RHOXF2 Antibody (PCRP-RHOXF2-1D7), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308529AF488
Description
RHOXF2 Monoclonal antibody specifically detects RHOXF2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Protein ArraySpecifications
RHOXF2 | |
Monoclonal | |
Alexa Fluor 488 | |
CT107, Paired-like homeobox protein PEPP-2, PEPP subfamily gene 2, PEPP-2, PEPP2cancer/testis antigen 107, rhox homeobox family member 2, Rhox homeobox family, member 2, Testis homeobox gene 1, THG1homeobox protein from AL590526 | |
Recombinant full-length human RHOXF2 protein (Uniprot: Q9BQY4 ) | |
100 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunofluorescence, Protein Array | |
PCRP-RHOXF2-1D7 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
84528 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction