Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RHOXF2 Monoclonal Antibody (3C4)
GREENER_CHOICE

Catalog No. PIMA522766
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA522766 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA522766 Supplier Invitrogen™ Supplier No. MA522766
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MEPPDQCSQY MTSLLSPAVD DEKELQDMNA MVLSLTEEVK EEEEDAQPEP EQGTAAGEKL KSAGAQGGEE KDGGGEEKDG GGAGVPGHLW EGDLEGTSGS DGNVEDSDQS EKEPGQQYSR PRGAVGGLEP GNAQQPNVHA FTPLQLQELE RIFQREQFPS EFLRRRLARS MNVTELAVQI WFENRRAKWR RHQRALMARN MLPFMAVGQP VMVTAAEAIT APLFISGMRD DYFWDHSHSS SLCFPMPPFP PPSLPLPLML LPPMPPAGQA EFGPFPFVIV PSFTFPNV.

This gene, which encodes a transcriptional repressor, is one of two paralogous X-linked homeobox-containing genes and is highly expressed in a variety of cancers. In addition, the encoded protein associates with the cell membrane and with microtubules, and is concentrated at the leading edge of migratory cells., Dec 2015].
TRUSTED_SUSTAINABILITY

Specifications

Antigen RHOXF2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3C4
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene RHOXF2
Gene Accession No. Q9BQY4
Gene Alias cancer/testis antigen 107; CT107; homeobox protein from AL590526; paired-like homeobox protein PEPP-2; PEPP subfamily gene 2; PEPP2; PEPP-2; Rhox homeobox family member 2; Rhox homeobox family, member 2; RHOXF2; Testis homeobox gene 1; THG1
Gene Symbols RHOXF2
Host Species Mouse
Immunogen RHOXF2 (AAH21719.1, 1 a.a. ~ 288 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84528
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.