Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RHOXF2 Mouse anti-Human, Clone: PCRP-RHOXF2-1D7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30740120UG
Description
RHOXF2 Monoclonal specifically detects RHOXF2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Protein Array.Specifications
RHOXF2 | |
Monoclonal | |
0.2 mg/ml | |
Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array | |
CT107, Paired-like homeobox protein PEPP-2, PEPP subfamily gene 2, PEPP-2, PEPP2cancer/testis antigen 107, rhox homeobox family member 2, Rhox homeobox family, member 2, Testis homeobox gene 1, THG1homeobox protein from AL590526 | |
Recombinant full-length human RHOXF2 protein | |
20 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunofluorescence, Peptide Array | |
PCRP-RHOXF2-1D7 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
84528 | |
Store at 4°C. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction