Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RHOXF2 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501728479 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-8479 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-8479 Supplier Proteintech Group Inc Supplier No. 130881AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene, which encodes a transcriptional repressor, is one of two paralogous X-linked homeobox-containing genes and is highly expressed in a variety of cancers. In addition, the encoded protein associates with the cell membrane and with microtubules, and is concentrated at the leading edge of migratory cells., Dec 2015]

Specifications

Antigen RHOXF2
Applications Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene RHOXF2
Gene Accession No. Q9BQY4
Gene Alias PEPP 2, PEPP2, Rhox homeobox family member 2, Rhox homeobox family, member 2, RHOXF2, Testis homeobox gene 1, THG1
Gene Symbols RHOXF2
Host Species Rabbit
Immunogen RHOXF2 Fusion Protein Ag3602
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84528
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.