Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RHPN1-AS1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144181
Description
RHPN1-AS1 Polyclonal Antibody for IHC (P), ELISA
RHPN1-AS1, also known as RHPN1 antisense RNA 1, is a long non-coding RNA (lncRNA) molecule. It is transcribed from the opposite strand of the RHPN1 gene, which encodes a protein called rhophilin 1. Long non-coding RNAs are RNA molecules that are longer than 200 nucleotides and do not code for proteins. They have been found to play important roles in various cellular processes, including gene regulation, chromatin remodeling, and cellular development. The specific function of RHPN1-AS1 is not yet fully understood, and further research is required to elucidate its role in cellular processes and its potential implications for human health and disease.
Specifications
RHPN1-AS1 | |
Polyclonal | |
Unconjugated | |
RHPN1-AS1 | |
C8orf51 | |
Rabbit | |
Protein G | |
RUO | |
78998 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin) | |
4 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
Q9BWJ2 | |
RHPN1-AS1 | |
Recombinant Human Putative uncharacterized protein encoded by RHPN1-AS1 protein (1-59AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction