Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RHPN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103248
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103248 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103248 Supplier Invitrogen™ Supplier No. PA5103248
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total RHPN1.

Has no enzymatic activity. May serve as a target for Rho, and interact with some cytoskeletal component upon Rho binding or relay a Rho signal to other molecules.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RHPN1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RHPN1
Gene Accession No. Q8TCX5
Gene Alias BB023497; Grbp; GTP-rho binding protein 1; GTP-Rho-binding protein 1; KIAA1929; mKIAA1929; ODF5; outer dense fiber of sperm tails 5; RHOPHILIN; rhophilin 1; rhophilin Rho GTPase binding protein 1; rhophilin, Rho GTPase binding protein 1; rhophilin-1; RHPN; RHPN1
Gene Symbols RHPN1
Host Species Rabbit
Immunogen A synthesized peptide derived from human RHPN1(Accession Q8TCX5), corresponding to amino acid residues 651-665.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114822
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.