Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RHPN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555039
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555039 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555039 Supplier Invitrogen™ Supplier No. PA555039
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EVVTELKAAG EAGASLQVVS LLPSSRLPSL GDRRPVLLGP RGLLRSQREH GCKTPASTWA SPRPLLNWSR KAQQGKTGGC PQPCAPVKPA P Highest antigen sequence identity to the following orthologs: Mouse - 49%, Rat - 46%.

Has no enzymatic activity. May serve as a target for Rho, and interact with some cytoskeletal component upon Rho binding or relay a Rho signal to other molecules.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RHPN1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RHPN1
Gene Accession No. Q8TCX5
Gene Alias BB023497; Grbp; GTP-rho binding protein 1; GTP-Rho-binding protein 1; KIAA1929; mKIAA1929; ODF5; outer dense fiber of sperm tails 5; RHOPHILIN; rhophilin 1; rhophilin Rho GTPase binding protein 1; rhophilin, Rho GTPase binding protein 1; rhophilin-1; RHPN; RHPN1
Gene Symbols RHPN1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RHPN1. Recombinant protein control fragment (Product #RP-93852).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114822
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.