Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RHPN1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555149
Description
Immunogen sequence: AFSLLRENFS HAPSPDMSAA SLCALEQLMM AQAQECVFEG LSPPASMAPQ DCLAQLRLAQ EAAQVAAEYR LVHRTMAQPP VHDYVPVSWT ALVHVKAE Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 73%.
Has no enzymatic activity. May serve as a target for Rho, and interact with some cytoskeletal component upon Rho binding or relay a Rho signal to other molecules.
Specifications
| RHPN1 | |
| Polyclonal | |
| Unconjugated | |
| RHPN1 | |
| BB023497; Grbp; GTP-rho binding protein 1; GTP-Rho-binding protein 1; KIAA1929; mKIAA1929; ODF5; outer dense fiber of sperm tails 5; RHOPHILIN; rhophilin 1; rhophilin Rho GTPase binding protein 1; rhophilin, Rho GTPase binding protein 1; rhophilin-1; RHPN; RHPN1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 114822 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.6 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TCX5 | |
| RHPN1 | |
| Recombinant protein corresponding to Human RHPN1. Recombinant protein control fragment (Product #RP-93853). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction