Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RHPN2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562469
Description
Immunogen sequence: DHQEKCLSQL YDHMPEGLTP LATLKNDQQR RQLGKSHLRR AMAHHEESVR EASLCKKLRS IEVLQKVLCA AQERSRLTYA QHQEEDDLLN L Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 71%.
This gene encodes a member of the rhophilin family of Ras-homologous (Rho)-GTPase binding proteins. The encoded protein binds both GTP- and GDP-bound RhoA and GTP-bound RhoB and may be involved in the organization of the actin cytoskeleton.
Specifications
RHPN2 | |
Polyclonal | |
Unconjugated | |
RHPN2 | |
1300002E07Rik; 76 kDa RhoB effector protein; AA536890; C85860; D7Ertd784e; GTP-Rho-binding protein 2; p76RBE; RHOBP; rhophilin 2; rhophilin Rho GTPase binding protein 2; rhophilin, Rho GTPase binding protein 2; rhophilin-2; rhophilin-like Rho-GTPase binding protein; RHPN2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
85415 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IUC4 | |
RHPN2 | |
Recombinant protein corresponding to Human RHPN2. Recombinant protein control fragment (Product #RP-99090). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction