Learn More
RHPN2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 126711AP150UL

Description
The MW of this protein is 77 kDa, and this antibody specifically recognizes the 77 kDa protein.
This gene encodes a member of the rhophilin family of Ras-homologous (Rho)-GTPase binding proteins. The encoded protein binds both GTP- and GDP-bound RhoA and GTP-bound RhoB and may be involved in the organization of the actin cytoskeleton.Specifications
RHPN2 | |
Polyclonal | |
Unconjugated | |
RHPN2 | |
76 kDa RhoB effector protein, GTP Rho binding protein 2, p76RBE, RhoBP, Rhophilin 2, RHPN2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
308516, 52428, 85415 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8BWR8, Q8IUC4 | |
Rhpn2 | |
RHPN2 Fusion Protein Ag3353 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.